Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethylcyclohexanecarboxylic acid (cis and trans mixture) 97%

Supplier: AMBEED, INC
Warning

A168568-100G A168568-10G A168568-1G A168568-250MG A168568-25G A168568-5G
77286-708EA 58 USD InStock
77286-708 77286-710 77286-712 77286-714 77286-716 77286-718
4-Ethylcyclohexanecarboxylic acid (cis and trans mixture) 97%
4-Ethylcyclohexanecarboxylic acid (cis and trans mixture)
Precaución: for Research Use Only


Formula: C₉H₁₆O₂
MDL Number: MFCD01568857
CAS Number: 91328-77-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty