Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-p-toluic acid 98%

Supplier: AMBEED, INC
Warning

A168805-100G A168805-10G A168805-25G A168805-500G
77273-630EA 19 USD InStock
77273-630 77273-632 77273-634 77748-125
3-Amino-p-toluic acid 98%
3-Amino-p-toluic acid
Precaución: for Research Use Only


Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 363
Melting Pt: 166
MDL Number: MFCD00007742
CAS Number: 2458-12-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty