Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methanesulfonato{[4-(N,N-dimethylamino)phenyl]di-t-butylphosphino}(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98%

Supplier: AMBEED, INC
Warning

A169489-1G A169489-250MG
77247-936EA 107 USD InStock
77247-936 77615-418
Methanesulfonato{[4-(N,N-dimethylamino)phenyl]di-t-butylphosphino}(2'-amino-1,1'-biphenyl-2-yl)palladium(II) 98%
Methanesulfonato{[4-(N,N-dimethylamino)phenyl]di-t-butylphosphino}(2'-amino-1,1'-biphenyl-2-yl)palladium(II)
Precaución: for Research Use Only


Formula: C₂₉H₄₁N₂O₃PPdS
MDL Number: MFCD27978423
CAS Number: 1820817-64-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty