Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-3-fluorophenol hydrochloride 96%

Supplier: AMBEED, INC
Warning

A169818-10G A169818-1G A169818-25G
77259-904EA 40 USD InStock
77259-904 77259-906 77259-908
4-Amino-3-fluorophenol hydrochloride 96%
4-Amino-3-fluorophenol hydrochloride
Precaución: for Research Use Only


Formula: C₆H₇ClFNO
MDL Number: MFCD01632189
CAS Number: 18266-53-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty