Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dimethyl-4-hydroxybenzonitrile 98%

Supplier: AMBEED, INC
Warning

A170112-100G A170112-1G A170112-25G A170112-5G A170112-10G
77286-792EA 175 USD InStock
77286-792 77286-794 77286-796 77286-798 77748-128
2,6-Dimethyl-4-hydroxybenzonitrile 98%
2,6-Dimethyl-4-hydroxybenzonitrile
Precaución: for Research Use Only


Formula: C₉H₉NO
MDL Number: MFCD00238591
CAS Number: 58537-99-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty