Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Luteolin-7-glucoside 98%

Supplier: AMBEED, INC
Warning

A170248-1MG A170248-5MG A170248-10MG
77236-254EA 30 USD InStock
77236-254 77236-256 77746-133
Luteolin-7-glucoside 98%
Luteolin-7-glucoside
Precaución: for Research Use Only


Formula: C₂₁H₂₀O₁₁
MW: 448.38 g/mol
MDL Number: MFCD06799436
CAS Number: 5373-11-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty