Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Bromomethyl)isoquinoline 95%

Supplier: AMBEED, INC
Danger

A172262-100MG A172262-1G A172262-250MG
77236-306EA 58 USD InStock
77236-306 77236-308 77236-310
1-(Bromomethyl)isoquinoline 95%
1-(Bromomethyl)isoquinoline
Precaución: for Research Use Only


Formula: C₁₀H₈BrN
MDL Number: MFCD06657041
CAS Number: 74417-44-0
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty