Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-6-methoxypyrimidin-2-amine 97%

Supplier: AMBEED, INC
Warning

A173062-10G A173062-25G A173062-500G A173062-100G
77273-782EA 5 USD InStock
77273-782 77273-784 77273-786 77748-139
4-Chloro-6-methoxypyrimidin-2-amine 97%
4-Chloro-6-methoxypyrimidin-2-amine
Precaución: for Research Use Only


Formula: C₅H₆ClN₃O
MW: 159.57 g/mol
MDL Number: MFCD00233535
CAS Number: 5734-64-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty