Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans,trans-4'-Propyl-4-(p-tolyl)bicyclohexyl 97%

Supplier: AMBEED, INC
Warning

A173244-10G A173244-1G A173244-25G A173244-5G
77286-930EA 22 USD InStock
77286-930 77286-932 77286-934 77286-936
trans,trans-4'-Propyl-4-(p-tolyl)bicyclohexyl 97%
trans,trans-4'-Propyl-4-(p-tolyl)bicyclohexyl
Precaución: for Research Use Only


Formula: C₂₂H₃₄
MDL Number: MFCD13182311
CAS Number: 84656-75-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty