Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

TP-0903 98%

Supplier: AMBEED, INC
Warning

A173278-1MG A173278-2MG
77236-322EA 32 USD InStock
77236-322 77236-324
TP-0903 98%
TP-0903
Precaución: for Research Use Only


Formula: C₂₄H₃₀ClN₇O₂S
MW: 516.06 g/mol
MDL Number: MFCD28502172
CAS Number: 1341200-45-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty