Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-nitrophenol 98%

Supplier: AMBEED, INC
Danger

A173501-100G A173501-10G A173501-25G A173501-500G A173501-5G
77286-938EA 42 USD InStock
77286-938 77286-940 77286-942 77286-944 77286-946
2-Fluoro-4-nitrophenol 98%
2-Fluoro-4-nitrophenol
Precaución: for Research Use Only


Formula: C₆H₄FNO₃
MDL Number: MFCD00051970
CAS Number: 403-19-0
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty