Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dioctyl carbonate 97%

Supplier: AMBEED, INC
Warning

A173641-100G A173641-500G
77260-006EA 22 USD InStock
77260-006 77260-008
Dioctyl carbonate 97%
Dioctyl carbonate
Precaución: For research use only.


Formula: C₁₇H₃₄O₃
MDL Number: MFCD00068968
CAS Number: 1680-31-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty