Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-5-(ethylthio)-2-methoxybenzoic acid 97%

Supplier: AMBEED, INC
Warning

A174619-10G A174619-25G A174619-5G A174619-100G
77286-968EA 11 USD InStock
77286-968 77286-970 77286-972 77748-142
4-Amino-5-(ethylthio)-2-methoxybenzoic acid 97%
4-Amino-5-(ethylthio)-2-methoxybenzoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₃NO₃S
MDL Number: MFCD04973620
CAS Number: 71675-86-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty