Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dicarboxyphenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A174621-100G A174621-1G A174621-25G A174621-5G A174621-250MG
77273-824EA 246 USD InStock
77273-824 77273-826 77273-828 77273-830 77748-143
3,5-Dicarboxyphenylboronic acid 97%
3,5-Dicarboxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₇BO₆
MDL Number: MFCD06203347
CAS Number: 881302-73-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty