Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2,5-Dichlorophenyl)propanoic acid 97%

Supplier: AMBEED, INC
Warning

A174729-100MG A174729-250MG A174729-1G
77248-124EA 12 USD InStock
77248-124 77248-126 77746-150
3-(2,5-Dichlorophenyl)propanoic acid 97%
3-(2,5-Dichlorophenyl)propanoic acid
Precaución: for Research Use Only


Formula: C₉H₈Cl₂O₂
MW: 219.07 g/mol
MDL Number: MFCD06823958
CAS Number: 68034-76-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty