Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4,5-Trifluorophenylboronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A175718-10G A175718-1G A175718-5G
77273-866EA 45 USD InStock
77273-866 77273-868 77273-870
3,4,5-Trifluorophenylboronic acid pinacol ester 97%
3,4,5-Trifluorophenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₂H₁₄BF₃O₂
MW: 258.0446 g/mol
MDL Number: MFCD05663885
CAS Number: 827614-70-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty