Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-3-(3-methoxyphenyl)propionic acid 95%

Supplier: AMBEED, INC
Warning

A175935-1G A175935-250MG A175935-5G
77273-872EA 35 USD InStock
77273-872 77273-874 77273-876
3-Amino-3-(3-methoxyphenyl)propionic acid 95%
3-Amino-3-(3-methoxyphenyl)propionic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₃NO₃
MDL Number: MFCD00462569
CAS Number: 68208-19-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty