Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4',4''-(2-Bromoethene-1,1,2-triyl)tris(bromobenzene) 97%

Supplier: AMBEED, INC
Warning

A177857-100MG A177857-250MG
77248-180EA 7 USD InStock
77248-180 77248-182
4,4',4''-(2-Bromoethene-1,1,2-triyl)tris(bromobenzene) 97%
4,4',4''-(2-Bromoethene-1,1,2-triyl)tris(bromobenzene)
Precaución: for Research Use Only


Formula: C₇H₁₃Cl₃Si
MDL Number: MFCD30609578
CAS Number: 408327-92-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty