Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-Hydroxyacetophenone 98%

Supplier: AMBEED, INC
Warning

A178509-1000G A178509-100G A178509-1MG A178509-500G A178509-25G
77287-070EA 51 USD InStock
77287-070 77287-072 77287-074 77287-076 77301-560
4'-Hydroxyacetophenone 98%
4'-Hydroxyacetophenone
Precaución: for Research Use Only


Formula: C₈H₈O₂
MDL Number: MFCD00002359
CAS Number: 99-93-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty