Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(2,6-difluoro-3-(1-hydropyrrol-1-yl)phenyl)titanocene 98%

Supplier: AMBEED, INC
Danger

A181017-25G A181017-5G
77260-232EA 95 USD InStock
77260-232 77260-234
Bis(2,6-difluoro-3-(1-hydropyrrol-1-yl)phenyl)titanocene 98%
Bis(2,6-difluoro-3-(1-hydropyrrol-1-yl)phenyl)titanocene
Precaución: for Research Use Only


Formula: C₃₀H₂₂F₄N₂Ti
MDL Number: MFCD00274256
CAS Number: 125051-32-3
UN: 1325
ADR: 4.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty