Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-iodoanisole 95%

Supplier: AMBEED, INC
Warning

A181339-100MG A181339-10G A181339-1G A181339-250MG A181339-5G
77287-228EA 7 USD InStock
77287-228 77287-230 77287-232 77287-234 77287-236
4-Bromo-3-iodoanisole 95%
4-Bromo-3-iodoanisole
Precaución: for Research Use Only


Formula: C₇H₆BrIO
MDL Number: MFCD00070743
CAS Number: 4897-68-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty