Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Butane-1,3-diyldibenzene 97%

Supplier: AMBEED, INC
Warning

A181832-250MG A181832-25G A181832-5G
77236-592EA 88 USD InStock
77236-592 77236-594 77236-596
Butane-1,3-diyldibenzene 97%
Butane-1,3-diyldibenzene
Precaución: For research use only.


Formula: C₁₆H₁₈
MDL Number: MFCD00506195
CAS Number: 1520-44-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty