Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3-methoxypropane 98%

Supplier: AMBEED, INC
Danger

A182492-10G A182492-25G A182492-100G
77236-612EA 5 USD InStock
77236-612 77236-614 77746-186
1-Bromo-3-methoxypropane 98%
1-Bromo-3-methoxypropane
Precaución: for Research Use Only


Formula: C₄H₉BrO
MW: 153.02 g/mol
MDL Number: MFCD02258473
CAS Number: 36865-41-5
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty