Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-methylnicotinate 97%

Supplier: AMBEED, INC
Warning

A183349-100G A183349-1G A183349-25G A183349-500G A183349-5G
77248-336EA 191 USD InStock
77248-336 77248-338 77248-340 77248-342 77248-344
Methyl-2-methylnicotinate 97%
Methyl-2-methylnicotinate
Precaución: for Research Use Only


Formula: C₈H₉NO₂
MDL Number: MFCD00051961
CAS Number: 65719-09-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty