Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

HMN-154 97%

Supplier: AMBEED, INC
Warning

A183563-10MG A183563-1MG A183563-25MG A183563-50MG A183563-5MG
77260-296EA 303 USD InStock
77260-296 77260-298 77260-300 77260-302 77260-304
HMN-154 97%
HMN-154
Precaución: for Research Use Only


Formula: C₂₀H₁₈N₂O₃S
MW: 366.43 g/mol
MDL Number: MFCD00945104
CAS Number: 173528-92-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty