Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-chloro-3-formylbenzoate 95%

Supplier: AMBEED, INC
Warning

A183908-100MG A183908-1G A183908-250MG A183908-5G
77260-320EA 18 USD InStock
77260-320 77260-322 77260-324 77260-326
Methyl 4-chloro-3-formylbenzoate 95%
Methyl 4-chloro-3-formylbenzoate
Precaución: for Research Use Only


Formula: C₉H₇ClO₃
MDL Number: MFCD16036863
CAS Number: 1044920-98-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty