Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5H-Benzofuro[3,2-c]carbazole 97%

Supplier: AMBEED, INC
Warning

A183982-10G A183982-1G A183982-25G A183982-5G
77260-328EA 127 USD InStock
77260-328 77260-330 77260-332 77260-334
5H-Benzofuro[3,2-c]carbazole 97%
5H-Benzofuro[3,2-c]carbazole
Precaución: for Research Use Only


Formula: C₁₈H₁₁NO
MDL Number: MFCD28129442
CAS Number: 1199616-66-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty