Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-N-amido-PEG9-acid 98%

Supplier: AMBEED, INC
Warning

A184757-100MG A184757-1G A184757-250MG A184757-5G
77248-384EA 21 USD InStock
77248-384 77248-386 77248-388 77746-193
Fmoc-N-amido-PEG9-acid 98%
Fmoc-N-amido-PEG9-acid
Precaución: for Research Use Only


Formula: C₃₆H₅₃NO₁₃
MW: 707.805 g/mol
MDL Number: MFCD28385470
CAS Number: 1191064-81-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty