Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dimethyluracil 97%

Supplier: AMBEED, INC
Warning

A184839-100G A184839-25G A184839-1000G
77274-240EA 49 USD InStock
77274-240 77274-242 77274-244 77274-246 77274-248 77748-201
1,3-Dimethyluracil 97%
1,3-Dimethyluracil
Precaución: for Research Use Only


Formula: C₆H₈N₂O₂
MDL Number: MFCD00038065
CAS Number: 874-14-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty