Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Benzyloxy-2-chloropyrimidine 97%

Supplier: AMBEED, INC
Warning

A185304-100MG A185304-1G A185304-250MG A185304-5G
77287-384EA 27 USD InStock
77287-384 77287-386 77287-388 77287-390
4-Benzyloxy-2-chloropyrimidine 97%
4-Benzyloxy-2-chloropyrimidine
Precaución: for Research Use Only


Formula: C₁₁H₉ClN₂O
MW: 220.66 g/mol
MDL Number: MFCD06798233
CAS Number: 108381-28-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty