Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenyl-4-(trifluoromethyl)-1H-imidazole 97%

Supplier: AMBEED, INC
Warning

A186106-1G A186106-250MG
77274-296EA 74 USD InStock
77274-296 77274-298
2-Phenyl-4-(trifluoromethyl)-1H-imidazole 97%
2-Phenyl-4-(trifluoromethyl)-1H-imidazole
Precaución: for Research Use Only


Formula: C₁₀H₇F₃N₂
MDL Number: MFCD10000861
CAS Number: 33469-36-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty