Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl potassium oxalate 97%

Supplier: AMBEED, INC
Danger

A186213-10G A186213-1G A186213-250MG A186213-25G A186213-5G
77260-384EA 134 USD InStock
77260-384 77260-386 77260-388 77260-390 77260-392
Methyl potassium oxalate 97%
Methyl potassium oxalate
Precaución: for Research Use Only


Formula: C₃H₃O₄·K
MDL Number: MFCD20922789
CAS Number: 10304-09-3
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty