Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 2-(2-hydroxyethoxy)acetate 95%

Supplier: AMBEED, INC
Warning

A186948-100MG A186948-250MG
77236-702EA 52 USD InStock
77236-702 77236-704
Sodium 2-(2-hydroxyethoxy)acetate 95%
Sodium 2-(2-hydroxyethoxy)acetate
Precaución: For research use only.


Formula: C₄H₇NaO₄
MDL Number: MFCD27500884
CAS Number: 142047-97-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty