Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2-(trifluoromethyl)benzonitrile 98%

Supplier: AMBEED, INC
Warning

A187542-1000G A187542-100G A187542-10G A187542-1G A187542-25G A187542-500G A187542-5G
77274-332EA 191 USD InStock
77274-332 77274-334 77274-336 77274-338 77274-340 77274-342 77274-344
4-Amino-2-(trifluoromethyl)benzonitrile 98%
4-Amino-2-(trifluoromethyl)benzonitrile
Precaución: for Research Use Only


Formula: C₈H₅F₃N₂
MW: 186.14 g/mol
MDL Number: MFCD00042155
CAS Number: 654-70-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty