Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Chlorobutyl)-1,3-dioxolane 97%

Supplier: AMBEED, INC
Danger

A188385-10G A188385-1G A188385-25G A188385-5G
77236-778EA 62 USD InStock
77236-778 77236-780 77236-782 77236-784
2-(4-Chlorobutyl)-1,3-dioxolane 97%
2-(4-Chlorobutyl)-1,3-dioxolane
Precaución: for Research Use Only


Formula: C₆H₁₁ClO₂
MDL Number: MFCD00043136
CAS Number: 118336-86-0
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty