Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

LDN-212854 98%

Supplier: AMBEED, INC
Danger

A189348-1MG A189348-5MG
77236-834EA 56 USD InStock
77236-834 77236-836
LDN-212854 98%
LDN-212854
Precaución: for Research Use Only


Formula: C₂₅H₂₂N₆
MDL Number: MFCD28099808
CAS Number: 1432597-26-6
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty