Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Naphthoyl chloride 97%

Supplier: AMBEED, INC
Danger

A190438-25G A190438-5G A190438-100G
77236-864EA 32 USD InStock
77236-864 77236-866 77746-219
1-Naphthoyl chloride 97%
1-Naphthoyl chloride
Precaución: for Research Use Only


Formula: C₁₁H₇ClO
MDL Number: MFCD00004002
CAS Number: 879-18-5
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty