Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-tert-Butoxy carbonylaminophenyl acetic acid 98%

Supplier: AMBEED, INC
Warning

A191155-1G A191155-250MG A191155-5G
77274-432EA 112 USD InStock
77274-432 77274-434 77274-436
3-tert-Butoxy carbonylaminophenyl acetic acid 98%
3-tert-Butoxy carbonylaminophenyl acetic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₇NO₄
MDL Number: MFCD02682313
CAS Number: 123036-51-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty