Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-(trifluoromethyl)aniline 98%

Supplier: AMBEED, INC
Warning

A191489-100G A191489-10G A191489-1G A191489-25G A191489-500G A191489-5G
77274-438EA 40 USD InStock
77274-438 77274-440 77274-442 77274-444 77274-446 77274-448
4-Fluoro-3-(trifluoromethyl)aniline 98%
4-Fluoro-3-(trifluoromethyl)aniline
Precaución: for Research Use Only


Formula: C₇H₅F₄N
MDL Number: MFCD00007834
CAS Number: 2357-47-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty