Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L(+)-Citrulline 98%

Supplier: AMBEED, INC
Warning

A192054-1000G A192054-100G A192054-100MG A192054-10G A192054-25G
77274-456EA 82 USD InStock
77274-456 77274-458 77274-460 77274-462 77274-464
L(+)-Citrulline 98%
L(+)-Citrulline
Precaución: for Research Use Only


Formula: C₆H₁₃N₃O₃
MW: 175.19 g/mol
MDL Number: MFCD00064397
CAS Number: 372-75-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty