Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

KPT-276 98%

Supplier: AMBEED, INC
Warning

A192347-10MG A192347-1MG A192347-50MG A192347-5MG
77236-938EA 64 USD InStock
77236-938 77236-940 77236-942 77746-223
KPT-276 98%
KPT-276
Precaución: for Research Use Only


Formula: C₁₆H₁₀F₈N₄O
MDL Number: MFCD27937049
CAS Number: 1421919-75-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty