Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Phenylbenzyl bromide 97%

Supplier: AMBEED, INC
Danger

A193384-1G A193384-5G
77274-542EA 45 USD InStock
77274-542 77274-544
2-Phenylbenzyl bromide 97%
2-Phenylbenzyl bromide
Precaución: for Research Use Only


Formula: C₁₃H₁₁Br
MDL Number: MFCD00075454
CAS Number: 19853-09-9
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty