Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxyethylurea 95%

Supplier: AMBEED, INC
Warning

A193561-1000G A193561-100G A193561-10G A193561-25G A193561-500G
77260-558EA 127 USD InStock
77260-558 77260-560 77260-562 77260-564 77260-566
2-Hydroxyethylurea 95%
2-Hydroxyethylurea
Precaución: for Research Use Only


Formula: C₃H₈N₂O₂
MDL Number: MFCD00059080
CAS Number: 2078-71-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty