Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Bromophenyl)-2,6-diphenylpyrimidine 97%

Supplier: AMBEED, INC
Warning

A194113-1G A194113-25G A194113-5G
77248-626EA 5 USD InStock
77248-626 77248-628 77248-630
4-(4-Bromophenyl)-2,6-diphenylpyrimidine 97%
4-(4-Bromophenyl)-2,6-diphenylpyrimidine
Precaución: for Research Use Only


Formula: C₂₂H₁₅BrN₂
MDL Number: MFCD00474667
CAS Number: 58536-46-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty