Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethoxy-5-methylbenzeneboronic acid 97%

Supplier: AMBEED, INC
Warning

A194243-10G A194243-1G A194243-25G A194243-5G
77274-576EA 47 USD InStock
77274-576 77274-578 77274-580 77274-582 77274-584
2-Ethoxy-5-methylbenzeneboronic acid 97%
2-Ethoxy-5-methylbenzeneboronic acid
Precaución: for Research Use Only


Formula: C₉H₁₃BO₃
MDL Number: MFCD00196832
CAS Number: 123291-97-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty