Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-((4-Guanidinobenzoyl)oxy)phenyl)acetic acid methanesulfonic acid salt 98%

Supplier: AMBEED, INC
Warning

A195521-10MG A195521-1MG A195521-25MG A195521-5MG
77260-646EA 99 USD InStock
77260-646 77260-648 77260-650 77260-652
2-(4-((4-Guanidinobenzoyl)oxy)phenyl)acetic acid methanesulfonic acid salt 98%
2-(4-((4-Guanidinobenzoyl)oxy)phenyl)acetic acid methanesulfonic acid salt
Precaución: for Research Use Only


Formula: C₁₇H₁₉N₃O₇S
MW: 409.41 g/mol
MDL Number: MFCD09842546
CAS Number: 71079-09-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty