Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Butyl-1,3,2-dioxaborolane-4s,5s-dicarboxylicacidbis(dimethylamidE) 95%

Supplier: AMBEED, INC
Warning

A196064-1G A196064-250MG A196064-25G
77260-662EA 22 USD InStock
77260-662 77260-664 77260-666
2-Butyl-1,3,2-dioxaborolane-4s,5s-dicarboxylicacidbis(dimethylamidE) 95%
2-Butyl-1,3,2-dioxaborolane-4s,5s-dicarboxylicacidbis(dimethylamidE)
Precaución: for Research Use Only


Formula: C₁₂H₂₃BN₂O₄
MDL Number: MFCD05663838
CAS Number: 161344-84-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty