Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluorophenylboronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A196189-100G A196189-10G A196189-1G A196189-250MG A196189-25G A196189-5G
77260-670EA 157 USD InStock
77260-670 77260-672 77260-674 77260-676 77260-678 77260-680
2-Fluorophenylboronic acid pinacol ester 97%
2-Fluorophenylboronic acid pinacol ester


Formula: C₁₂H₁₆BFO₂
MDL Number: MFCD05663874
CAS Number: 876062-39-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty