Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dibromo-5-fluoropyridine 98%

Supplier: AMBEED, INC
Warning

A196198-100G A196198-10G A196198-1G A196198-25G A196198-5G
77248-706EA 207 USD InStock
77248-706 77248-708 77248-710 77248-712 77248-714
2,3-Dibromo-5-fluoropyridine 98%
2,3-Dibromo-5-fluoropyridine
Precaución: for Research Use Only


Formula: C₅H₂Br₂FN
MDL Number: MFCD08532501
CAS Number: 878207-82-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty