Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Methylhydroxylammonium chloride 97%

Supplier: AMBEED, INC
Warning

A196651-100G A196651-10G A196651-1G A196651-25G A196651-500G A196651-5G
77274-636EA 91 USD InStock
77274-636 77274-638 77274-640 77274-642 77274-644 77274-646
N-Methylhydroxylammonium chloride 97%
N-Methylhydroxylammonium chloride
Precaución: for Research Use Only


Formula: CH₆ClNO
MDL Number: MFCD00012597
CAS Number: 4229-44-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty